Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-nitrooleic acid | 875685-46-4 | MFCD12912274 | 98.9% | 327.46 g·mol-1 | C18H33NO4 | 500 UG
SDP

Catalog No. 5000245680
Encompass_Preferred

10-Nitrooleic acid is a nitro fatty acid research compound used to probe redox and inflammatory signaling pathways. It has reported effects in models of oxidative stress, inflammation, fibrosis, and cytoprotection. The material is supplied as a small-scale ethanol solution for laboratory research; consult the product datasheet and safety documentation for proper handling and storage.

  • Liquid, colorless to light yellow.
  • Molecular weight 327.46 g·mol-1.
  • Chemical formula C18H33NO4.
  • Supplied as 500 μg in 1 mL ethanol.
  • Purity 98.9% (as reported by supplier).
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-45680 Supplier Medchemexpress LLC Supplier No. HY101559500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

10-Nitrooleic acid is a nitro fatty acid research compound used to probe redox and inflammatory signaling pathways. It has reported effects in models of oxidative stress, inflammation, fibrosis, and cytoprotection. The material is supplied as a small-scale ethanol solution for laboratory research; consult the product datasheet and safety documentation for proper handling and storage.

  • Liquid, colorless to light yellow.
  • Molecular weight 327.46 g·mol-1.
  • Chemical formula C18H33NO4.
  • Supplied as 500 μg in 1 mL ethanol.
  • Purity 98.9% (as reported by supplier).
  • For research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.