Learn More
Seracare Life Sciences Inc 10% NORMAL MOUSE SERUM
Catalog No. 5067586
Encompass_Preferred
Normal Mouse Serum can be used as a blocking agent to treat plastic surfaces, membrane or tissue after they have been sensitized with primary antibody or antigen. It provides an alternative to bovine serum albumin (BSA) and non-fat dry milk. It is effective in reducing nonspecific binding of proteins to reaction surfaces, thereby maximizing signal-to-noise ratio. This blocking agent is recommended for use in immunoassays where the primary antibody was produced in mouse, as a source of non-specific serum protein or on mouse tissue for immunohistochemical applications.
This item is not returnable. View return policy
Catalog No. 50-675-86 Supplier Seracare Life Sciences Inc Supplier No. 55600010
Normal Mouse Serum can be used as a blocking agent to treat plastic surfaces, membrane or tissue after they have been sensitized with primary antibody or antigen. It provides an alternative to bovine serum albumin (BSA) and non-fat dry milk. It is effective in reducing nonspecific binding of proteins to reaction surfaces, thereby maximizing signal-to-noise ratio. This blocking agent is recommended for use in immunoassays where the primary antibody was produced in mouse, as a source of non-specific serum protein or on mouse tissue for immunohistochemical applications.