Learn More
Abcam 10% NP-40 Substitute, Detergent. 10% Solution in Water., 100ML
Supplier:  Abcam AB142227100ML
Encompass_Preferred
10% NP-40 detergent, substitute is a non-ionic detergent that isolates membrane-bound proteins under non-denaturing conditions. - Cited in over 35 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4931