Learn More
Chemglass Life Sciences Agitator, PTFE, Four Blade, 10in Diameter, Fits 25.4mm Shaft Size, 45° Impeller Angle
SDP
Catalog No. CG209522
Encompass_Preferred
Agitator, PTFE, Four Blade, 10in Diameter, Fits 25.4mm Shaft Size, 45¿ Impeller Angle. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 25.4mm OD stirrer shaft CG-2097 and have 45¿ impeller angles. The 45¿ impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. Agitator hub is open so that it can be positioned anywhere along the shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. PLEASE NOTE: ANGLED IMPELLARS DESIGNED TO STIR UPWARDS IN A CLOCKWISE DIRECTION ARE AVAILABLE. PLEASE CALL FOR PRICING.
Catalog No. CG209522 Supplier Chemglass Life Sciences Supplier No. CG209522
Agitator, PTFE, Four Blade, 10in Diameter, Fits 25.4mm Shaft Size, 45¿ Impeller Angle. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 25.4mm OD stirrer shaft CG-2097 and have 45¿ impeller angles. The 45¿ impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. Agitator hub is open so that it can be positioned anywhere along the shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. PLEASE NOTE: ANGLED IMPELLARS DESIGNED TO STIR UPWARDS IN A CLOCKWISE DIRECTION ARE AVAILABLE. PLEASE CALL FOR PRICING.