Learn More
AdipoGen 10-Octadecylacridine orange Br
SDP
Catalog No. 502560346
Encompass_Preferred
Chemical. CAS 75168-16-0. Formula C35H56BrN3. MW 598.74. Synthetic. Fluorescent reagent with high selectivity. Potential sensitive dye PSD to monitor differences in cell potential as they occur during ion transport through the cell membrane.
Catalog No. 50-256-0346 Supplier AdipoGen Supplier No. CDXO0018M250
Chemical. CAS 75168-16-0. Formula C35H56BrN3. MW 598.74. Synthetic. Fluorescent reagent with high selectivity. Potential sensitive dye PSD to monitor differences in cell potential as they occur during ion transport through the cell membrane.