Learn More
Cayman Chemical 10-PAHSA 13C4 100ug
SDP
Catalog No. 502994866
Encompass_Preferred
An internal standard for the quantification of 10-PAHSA by GC- or LC-MS
Catalog No. 50-299-4866 Supplier Cayman Chemical Supplier No. 22657100 UG
An internal standard for the quantification of 10-PAHSA by GC- or LC-MS