Learn More
Cayman Chemical 10-PAHSA-d31 100ug
SDP
Catalog No. 502995405
Encompass_Preferred
An internal standard for the quantification of 10-PAHSA by GC- or LC-MS
Catalog No. 50-299-5405 Supplier Cayman Chemical Supplier No. 22658100 UG
An internal standard for the quantification of 10-PAHSA by GC- or LC-MS