Learn More
Andwin Scientific 10-PANEL DRUG CARD TEST PK/25
SDP
Catalog No. 50465176
Encompass_Preferred
The 10-panel drug test card is used to quickly test urine in order to uncover the use of drugs, such as cocaine, amphetamine, methamphetamine, marijuana, opiate 2000, phencyclidine, barbiturate, bensodiazepine, and tricyclic antidepressants.. This product is sold in a pack of 25.
Catalog No. 50-465-176 Supplier Andwin Scientific Supplier No. 710422
The 10-panel drug test card is used to quickly test urine in order to uncover the use of drugs, such as cocaine, amphetamine, methamphetamine, marijuana, opiate 2000, phencyclidine, barbiturate, bensodiazepine, and tricyclic antidepressants.. This product is sold in a pack of 25.