Learn More
Newcomer Supply Paraformaldehyde 10%, Buffered, 1 Liter
SDP
Catalog No. NC1724168
Encompass
Use: Cell flow fixation. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC1724168 Supplier Newcomer Supply Supplier No. 13301B
Use: Cell flow fixation. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.