Learn More
Electron Microscopy Sciences 10% Paraformaldehyde (formaldehyde) aqueous solution, 10x10 mL
SDP
Catalog No. 50980490
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 10 mL ampoules sealed under inert gas.
Catalog No. 50-980-490 Supplier Electron Microscopy Sciences Supplier No. 15712
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 10 mL ampoules sealed under inert gas.