Learn More
Electron Microscopy Sciences 10% Paraformaldehyde (formaldehyde) aqueous solution, 100 mL
SDP
Catalog No. 50980491
Encompass_Preferred
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.
Catalog No. 50-980-491 Supplier Electron Microscopy Sciences Supplier No. 15712S
Paraformaldehyde - methanol free solution. A more efficient and rapid penetrant fixative used in combination with Glutaraldehyde and Acrolein fixatives. 100 ml bottles.