Learn More
DIAGNOX LLC 10 PARAMETER UA TESTS 30/PK
Catalog No. NC2866643
Encompass_Preferred
URINOX10-30 10 Parameter Urinalysis UA Reagent Strips (LEU,NIT, URO, BIL, PRO, PH, KET, GLU, BLO, SG). 30 per pack.CLIA Waived, FDA 510k Professional/Prescription Use, HealthCanada, CE, UKCA
Catalog No. NC2866643 Supplier DIAGNOX LLC Supplier No. URINOX1030
URINOX10-30 10 Parameter Urinalysis UA Reagent Strips (LEU,NIT, URO, BIL, PRO, PH, KET, GLU, BLO, SG). 30 per pack.CLIA Waived, FDA 510k Professional/Prescription Use, HealthCanada, CE, UKCA