Learn More
BTNX Inc 10 Parameter Urine Drug Screen - 25/kit (10.14) - COC300 AMP1000 MET1000 THC50 MTD300 OPI2000 OXY100 PCP25 BAR300 BZO300
Catalog No. NC3361249
Encompass_Preferred
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips. (10.14) - COC300 AMP1000 MET1000 THC50 MTD300 OPI2000 OXY100 PCP25 BAR300 BZO300
This item is not returnable. View return policy
Catalog No. NC3361249 Supplier BTNX Inc Supplier No. D10.141V
The Rapid Response™ Multi-Drug One Step Cup is an easy-to-use, one-step solution for drug screening that can simultaneously detect multiple drugs in urine samples. This format is a great alternative to the industry's complicated testing processes and is available with and without adulterant tests. It has a longer detection window than saliva tests and provides results in 5 minutes. This product is also available with an option of built-in Specimen Validity/Adulteration Test Strips. (10.14) - COC300 AMP1000 MET1000 THC50 MTD300 OPI2000 OXY100 PCP25 BAR300 BZO300