Learn More
JANT PHARMACAL CORPORATION ACCUSTRIP 10-Parameter Urine Reagent Strip for Use with ACCUSTRIP URS Reader 100 Tests/Vial
SDP
Catalog No. NC3457044
Encompass_Preferred
The Accustrip URS 10 Reagent Strip for Urinalysis is a dip-and-read test strip. The product is intended for use as an in vitro diagnostic aid using urine specimens to screen for diabetes, metabolic abnormalities, liver diseases, biliary and hepatic obstructions and diseases of the kidneys and urinary tract.The strip provides qualitative and semi-quantitative tests for blood, urobilinogen, bilirubin, protein, nitrite, ketones, glucose, pH, specific gravity and leukocytes by visual comparison with a color chart for each concentration range.The strips may be read visually, requiring no additional laboratory equipment. The test strip may also be read instrumentally using the Accustrip URS Reader.
This item is not returnable. View return policy
Catalog No. NC3457044 Supplier JANT PHARMACAL CORPORATION Supplier No. UA870
The Accustrip URS 10 Reagent Strip for Urinalysis is a dip-and-read test strip. The product is intended for use as an in vitro diagnostic aid using urine specimens to screen for diabetes, metabolic abnormalities, liver diseases, biliary and hepatic obstructions and diseases of the kidneys and urinary tract.The strip provides qualitative and semi-quantitative tests for blood, urobilinogen, bilirubin, protein, nitrite, ketones, glucose, pH, specific gravity and leukocytes by visual comparison with a color chart for each concentration range.The strips may be read visually, requiring no additional laboratory equipment. The test strip may also be read instrumentally using the Accustrip URS Reader.