Learn More
Teco Diagnostics 10 PARAMETER URINE STRIP
Catalog No. NC0343093
Encompass_Preferred
Teco Urine Reagent Strips (URS) for Urinalysis are firm plastic strips to which several different reagent areas are affixed. Depending on the product being used, Teco Urine Reagent Strips provide tests for Glucose, Bilirubin, Ketone (Acetoacetic acid), Specific Gravity, Blood, pH, Protein, Urobilin. 1x100 Strips Bottle, 1x Teco URS-10 Color Chart, Package Insert
Catalog No. NC0343093 Supplier Teco Diagnostics Supplier No. URS10100U
Teco Urine Reagent Strips (URS) for Urinalysis are firm plastic strips to which several different reagent areas are affixed. Depending on the product being used, Teco Urine Reagent Strips provide tests for Glucose, Bilirubin, Ketone (Acetoacetic acid), Specific Gravity, Blood, pH, Protein, Urobilin. 1x100 Strips Bottle, 1x Teco URS-10 Color Chart, Package Insert