Learn More
CLEAR BALLISTICS LLC 10% Gel FBI Block 6" x 6" x 16"
SDP
Catalog No. NC3432057
Encompass_Preferred
FBI Block is perfect for terminal ballistics testing for all handguns and rifles where penetrations are less than 16 inches. The FBI Block meets FBI ballistics testing protocol 100%, and completely simulates the testing performed by the FBI.
Catalog No. NC3432057 Supplier CLEAR BALLISTICS LLC Supplier No. 852844007000
FBI Block is perfect for terminal ballistics testing for all handguns and rifles where penetrations are less than 16 inches. The FBI Block meets FBI ballistics testing protocol 100%, and completely simulates the testing performed by the FBI.