Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready America™ 10 Person Emergency Kit
SDP

Catalog No. 19777021
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-777-021 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-777-021 Supplier Ready America™ Supplier No. 70551
  • $496.00
    $381.65 / Each of 1

    Save $114.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready America™ 10 Person Emergency Kit provides the food, water, sanitation, emergency lighting and first aid supplies for up to 10 people.

  • Includes the following:
    • 10 x 2,400 Calorie Food Bars (5-year shelf life)
    • 60 Water Packets 4.225 oz. (5-year shelf life)
    • 10 Emergency Lightsticks (12-hr)
    • 10 Emergency Survival Blankets
    • 10 Emergency Whistles
    • 20 Nitrile Gloves
    • 10 Dust Masks
    • 10 Pocket Tissue Packs
    • 5 Bio-Hazard Bags
    • 30 Pre- Moistened Towelettes
    • 10 Toothbrushes
    • 10 Toothpastes
    • 10 Mouthwashes
    • A 107-Piece All-Purpose First Aid Kit
    • A 12 Gallon Tote

Specifications

Product Type Emergency Supplies
Color Black
Material Plastic
Height (English) 12 in.
Height (Metric) 30.48 cm
Length (English) 20 in.
Length (Metric) 50.8 cm
Weight (English) 12 lb.
Weight (Metric) 5.45 kg
Width (English) 12 in.
Width (Metric) 30.48 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.