Learn More
Bulbman Inc ESA / FHD 64225 Osram Photo Optic 54260 Light Bulb, 10/pk
SDP
Catalog No. NC1768958
Encompass_Preferred
ESA / FHD 64225 Osram Photo Optic 54260 Light Bulb: 6 Volt, 10 Watt, T3 tungsten halogen lamp with a single ended G4 bi pin base.
Catalog No. NC1768958 Supplier Bulbman Inc Supplier No. 10361710
ESA / FHD 64225 Osram Photo Optic 54260 Light Bulb: 6 Volt, 10 Watt, T3 tungsten halogen lamp with a single ended G4 bi pin base.