Learn More
Medicus Health 10 Place, Stainless Steel Staining Rack with Handle
SDP
Catalog No. NC3136583
Encompass
10 Place Stainless Steel Slide Dipper is durable, resistant to chemicals and corrosion, and easily cleaned ∙ Holds standard 3"L x 1"W slides vertically ∙ 10 Place Stainless Steel Slide Dipper great for histological, cytological and hematological staining procedures with total immersion for slides∙ Great Hematology Departments who need to do Wright's Stains∙ Slide Staining Dipper is sold by the each∙ Slide Dipper measures: 1″W x 1.4″D x 3.4″H∙ Stainless Steel Staining Dipper is designed for 75mm x 25mm or 3" x 1" slides∙ Material: Stainless steel
Catalog No. NC3136583 Supplier Medicus Health Supplier No. 5441M2
10 Place Stainless Steel Slide Dipper is durable, resistant to chemicals and corrosion, and easily cleaned ∙ Holds standard 3"L x 1"W slides vertically ∙ 10 Place Stainless Steel Slide Dipper great for histological, cytological and hematological staining procedures with total immersion for slides∙ Great Hematology Departments who need to do Wright's Stains∙ Slide Staining Dipper is sold by the each∙ Slide Dipper measures: 1″W x 1.4″D x 3.4″H∙ Stainless Steel Staining Dipper is designed for 75mm x 25mm or 3" x 1" slides∙ Material: Stainless steel