Learn More
Fuelcellstore 10% Platinum Ruthenium on Vulcan, 1 g
SDP
Supplier:  Fuelcellstore 57080022
Encompass
10% Platinum Ruthenium on Vulcan XC 72 (Carbon) is most commonly used as the catalyst for methanol oxidation in Direct Methanol Fuel Cells (DMFC). This catalyst has a standard 1:1 atomic ratio.
Catalog No. NC1899233