Learn More
Vici Valco 10 PORT VALVES 0.25MM STATOR
SDP
Catalog No. NC3038583
Encompass_Preferred
10,000 psi, microbore, 1/16"
Catalog No. NC3038583 Supplier Vici Valco Supplier No. C721C70
10,000 psi, microbore, 1/16"