Learn More
Teknova 10% SDS Solution, 1000mL, Non-Sterile. 2 pack.
SDP
Catalog No. 502387436
Encompass
Sodium Dodecyl Sulfate (SDS) is an anionic surfactant solution and is one of the most commonly used detergents in protein purification and electrophoretic separation of proteins and lipids. It is often used as an adjuvant in lysing cells during DNA extraction and as a protein denaturing agent in SDS-PAGE.
Catalog No. 50-238-7436 Supplier Teknova Supplier No. S02892PK
Sodium Dodecyl Sulfate (SDS) is an anionic surfactant solution and is one of the most commonly used detergents in protein purification and electrophoretic separation of proteins and lipids. It is often used as an adjuvant in lysing cells during DNA extraction and as a protein denaturing agent in SDS-PAGE.