Learn More
Bio Rad Laboratories 10% SDS SOLUTION
Catalog No. NC9407597
Encompass
SDS solutions are prepared with 18 megaohm water and have no detectable DNase or RNase activity. This 10% (weight to volume) solution for electrophoresis applications is supplied as 250 ml. (161-0416)
Catalog No. NC9407597 Supplier Bio Rad Laboratories Supplier No. 1610416
SDS solutions are prepared with 18 megaohm water and have no detectable DNase or RNase activity. This 10% (weight to volume) solution for electrophoresis applications is supplied as 250 ml. (161-0416)