Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [10]-shogaol | 36752-54-2 | MFCD00916692 | 332.48 g/mol | C21H32O3 | 10 MG
SDP

Catalog No. 5000288961
Encompass_Preferred

[10]-Shogaol is a ginger-derived phenolic compound used as a research reagent with reported antioxidant, anti-inflammatory, and anticancer activities. The manufacturer datasheet includes physicochemical identifiers, solubility formulations, and storage guidance for in vitro and in vivo use.

  • Ginger-derived phenolic compound with bioactivity
  • Demonstrates antioxidant and anti-inflammatory effects
  • Suitable for in vitro and in vivo pharmacology studies
  • Soluble in DMSO; manufacturer provides vehicle formulations for in vivo dosing
  • Storage guidance: store at 4°C and protect from light; in solution store at -80°C or -20°C for defined stability
  • Available in small research quantities (e.g., 1 mg, 5 mg, 10 mg)

Catalog No. 50-002-88961 Supplier Medchemexpress LLC Supplier No. HYN243410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[10]-Shogaol is a ginger-derived phenolic compound used as a research reagent with reported antioxidant, anti-inflammatory, and anticancer activities. The manufacturer datasheet includes physicochemical identifiers, solubility formulations, and storage guidance for in vitro and in vivo use.

  • Ginger-derived phenolic compound with bioactivity
  • Demonstrates antioxidant and anti-inflammatory effects
  • Suitable for in vitro and in vivo pharmacology studies
  • Soluble in DMSO; manufacturer provides vehicle formulations for in vivo dosing
  • Storage guidance: store at 4°C and protect from light; in solution store at -80°C or -20°C for defined stability
  • Available in small research quantities (e.g., 1 mg, 5 mg, 10 mg)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.