Learn More
Andwin Scientific 10 SLIDE STAIN TRAY W/BLK LID
SDP
Catalog No. 50931028
Encompass_Preferred
The 10-Slide StainTray with Black Lid is constructed from tough ABS plastic that allows this product to be compatible with a wide range of chemicals (except chlorinated hydrocarbons). This product can take up to 20 slides on 4 plastic rails covered with a polymer strip to efficiently hold the slides even if the tray is held at an angle. The StainTray measures 9" x 9" x 1" (24 x 24 x 4.5 cm H) with the cover. The black cover is included with this product and is ideal for fluorescent work. This is sold individually.
Catalog No. 50-931-028 Supplier Andwin Scientific Supplier No. 71396B
The 10-Slide StainTray with Black Lid is constructed from tough ABS plastic that allows this product to be compatible with a wide range of chemicals (except chlorinated hydrocarbons). This product can take up to 20 slides on 4 plastic rails covered with a polymer strip to efficiently hold the slides even if the tray is held at an angle. The StainTray measures 9" x 9" x 1" (24 x 24 x 4.5 cm H) with the cover. The black cover is included with this product and is ideal for fluorescent work. This is sold individually.