Learn More
Teknova 10% Sodium Azide, 250mL, Sterile. 8 Pack.
SDP
Catalog No. 502745899
Encompass
Sodium azide is a versatile compound that has a wide range of applications in the laboratory. It can be used in dilute solutions as a preservative of biological samples and stock solutions, as well as an effective reagent for synthesis work.
Catalog No. 50-274-5899 Supplier Teknova Supplier No. 213348PK
Sodium azide is a versatile compound that has a wide range of applications in the laboratory. It can be used in dilute solutions as a preservative of biological samples and stock solutions, as well as an effective reagent for synthesis work.