Learn More
Teknova 10% Sodium Azide. 100mL, Sterile.
SDP
Catalog No. NC0080007
Encompass
Sodium azide is a versatile compound that has a wide range of applications in the laboratory. It can be used in dilute solutions as a preservative of biological samples and stock solutions, as well as an effective reagent for synthesis work.
This item is not returnable. View return policy
Catalog No. NC0080007 Supplier Teknova Supplier No. S0209
Sodium azide is a versatile compound that has a wide range of applications in the laboratory. It can be used in dilute solutions as a preservative of biological samples and stock solutions, as well as an effective reagent for synthesis work.