Learn More
Medchemexpress LLC 10-(tert-butoxy)-10-oxodecanoic acid | 234081-96-0 | MFCD28505598 | 98.0% | 258.35 g·mol⁻1 | C14H26O4 | 10 G
SDP
Catalog No. 5000299475
Encompass_Preferred
10-(tert-Butoxy)-10-oxodecanoic acid is a research-grade short linker featuring a carboxylic acid and a tert-butyl ester protecting group. The tert-butyl group can be selectively removed to reveal a free carboxylic acid for downstream coupling reactions, enabling synthesis of esters, amides, and conjugates. Supplied as a high-purity solid with solution options for convenient use.
  • Contains a carboxylic acid and a tert-butyl ester protecting group.
  • Tert-butyl group removable under standard deprotection conditions.
  • Suitable for esterification and amide coupling chemistry.
  • High purity supports synthetic and probe development workflows.
  • Available in multiple sizes and convenient DMSO solutions.
Catalog No. 50-002-99475 Supplier Medchemexpress LLC Supplier No. HYW03945810G
10-(tert-Butoxy)-10-oxodecanoic acid is a research-grade short linker featuring a carboxylic acid and a tert-butyl ester protecting group. The tert-butyl group can be selectively removed to reveal a free carboxylic acid for downstream coupling reactions, enabling synthesis of esters, amides, and conjugates. Supplied as a high-purity solid with solution options for convenient use.
  • Contains a carboxylic acid and a tert-butyl ester protecting group.
  • Tert-butyl group removable under standard deprotection conditions.
  • Suitable for esterification and amide coupling chemistry.
  • High purity supports synthetic and probe development workflows.
  • Available in multiple sizes and convenient DMSO solutions.