Learn More
Medchemexpress LLC 10-(tert-butoxy)-10-oxodecanoic acid | 234081-96-0 | MFCD28505598 | 98.0% | 258.35 g·mol⁻1 | C14H26O4 | 25 G
SDP
Catalog No. 5000304025
Encompass_Preferred
10-(tert-Butoxy)-10-oxodecanoic acid is a short organic linker containing a terminal carboxylic acid and a tert-butyl ester used as a building block in synthesis and biochemical assay development. It is provided as a high-purity research reagent, soluble in DMSO, and supplied in multiple laboratory pack sizes for convenient use.
  • Short linker with a terminal carboxylic acid.
  • Tert-butyl ester protecting group enables controlled deprotection.
  • High purity suitable for synthetic applications (98.0%).
  • Good solubility in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Stable storage as solid at room temperature for up to 3 years.
  • Available in multiple pack sizes for scalable experiments.
Catalog No. 50-003-04025 Supplier Medchemexpress LLC Supplier No. 25GHYW039458
10-(tert-Butoxy)-10-oxodecanoic acid is a short organic linker containing a terminal carboxylic acid and a tert-butyl ester used as a building block in synthesis and biochemical assay development. It is provided as a high-purity research reagent, soluble in DMSO, and supplied in multiple laboratory pack sizes for convenient use.
  • Short linker with a terminal carboxylic acid.
  • Tert-butyl ester protecting group enables controlled deprotection.
  • High purity suitable for synthetic applications (98.0%).
  • Good solubility in DMSO (100 mg/mL); ultrasonic agitation recommended.
  • Stable storage as solid at room temperature for up to 3 years.
  • Available in multiple pack sizes for scalable experiments.