Learn More
Medchemexpress LLC 10-(tert-butoxy)-10-oxodecanoic acid | 234081-96-0 | MFCD28505598 | ≥98.0% | 258.35 g·mol⁻¹ | C14H26O4 | 1 G
SDP
Catalog No. 5000301195
Encompass_Preferred
10-(tert-butoxy)-10-oxodecanoic acid is a small-molecule reagent that combines a terminal carboxylic acid with a tert-butoxy protected group. Supplied as a colorless to light yellow solid-liquid mixture with high purity, it is used as a short linker or building block in small-molecule synthesis, conjugation, and bioconjugation workflows.
  • Provides a terminal carboxylic acid functional group for conjugation and coupling reactions.
  • Contains a tert-butoxy protecting group to enable selective deprotection strategies.
  • High purity (≥98.0%) appropriate for research-grade synthesis and analytical work.
  • Supplied as a solid-liquid mixture, facilitating handling and formulation.
  • Available in multiple package sizes for scale flexibility.
Catalog No. 50-003-01195 Supplier Medchemexpress LLC Supplier No. HYW0394581G
10-(tert-butoxy)-10-oxodecanoic acid is a small-molecule reagent that combines a terminal carboxylic acid with a tert-butoxy protected group. Supplied as a colorless to light yellow solid-liquid mixture with high purity, it is used as a short linker or building block in small-molecule synthesis, conjugation, and bioconjugation workflows.
  • Provides a terminal carboxylic acid functional group for conjugation and coupling reactions.
  • Contains a tert-butoxy protecting group to enable selective deprotection strategies.
  • High purity (≥98.0%) appropriate for research-grade synthesis and analytical work.
  • Supplied as a solid-liquid mixture, facilitating handling and formulation.
  • Available in multiple package sizes for scale flexibility.