Learn More
Medchemexpress LLC 10-(tert-butoxy)-10-oxodecanoic acid | 234081-96-0 | MFCD28505598 | 98.0% | 258.35 g/mol | C14H26O4 | 5 G
SDP
Catalog No. 5000300532
Encompass_Preferred
10-(tert-Butoxy)-10-oxodecanoic acid is a tert-butyl-protected decanoic acid linker used in small-molecule synthesis and biochemical assays. The tert-butyl ester can be selectively removed to reveal the carboxylic acid for coupling, enabling preparation of esters or amides. Supplied for research use in solid or solution formats with supporting analytical documentation.
  • Tert-butyl protected carboxylic acid for selective deprotection.
  • Useful linker for forming esters and amides with alcohols or amines.
  • High purity suitable for research applications.
  • Available as solid or 10 mM solution in DMSO for flexibility.
  • Documented COA and SDS available for batch verification.
Catalog No. 50-003-00532 Supplier Medchemexpress LLC Supplier No. HYW0394585G
10-(tert-Butoxy)-10-oxodecanoic acid is a tert-butyl-protected decanoic acid linker used in small-molecule synthesis and biochemical assays. The tert-butyl ester can be selectively removed to reveal the carboxylic acid for coupling, enabling preparation of esters or amides. Supplied for research use in solid or solution formats with supporting analytical documentation.
  • Tert-butyl protected carboxylic acid for selective deprotection.
  • Useful linker for forming esters and amides with alcohols or amines.
  • High purity suitable for research applications.
  • Available as solid or 10 mM solution in DMSO for flexibility.
  • Documented COA and SDS available for batch verification.