Learn More
Medchemexpress LLC 10-((tert-butoxycarbonyl)amino)decanoic acid | 173606-50-3 | MFCD00270351 | >98.0% | 287.40 g·mol⁻¹ | C15H29NO4 | 25 G
SDP
Catalog No. 5000209024
Encompass_Preferred
Boc-10-aminodecanoic acid (10-((tert-butoxycarbonyl)amino)decanoic acid) is a Boc-protected amino acid used as a linker building block in peptide synthesis and PROTAC construction. It contains a terminal carboxylic acid and a Boc-protected amine that enable standard coupling reactions and selective deprotection.
  • Boc-protected amino acid with terminal carboxylic acid.
  • Suitable for peptide synthesis and PROTAC linker construction.
  • High purity for research applications.
  • Soluble in common organic solvents for coupling chemistry.
  • Available in multiple pack sizes, including 25 g.
Catalog No. 50-002-09024 Supplier Medchemexpress LLC Supplier No. 25GHYW089232
Boc-10-aminodecanoic acid (10-((tert-butoxycarbonyl)amino)decanoic acid) is a Boc-protected amino acid used as a linker building block in peptide synthesis and PROTAC construction. It contains a terminal carboxylic acid and a Boc-protected amine that enable standard coupling reactions and selective deprotection.
  • Boc-protected amino acid with terminal carboxylic acid.
  • Suitable for peptide synthesis and PROTAC linker construction.
  • High purity for research applications.
  • Soluble in common organic solvents for coupling chemistry.
  • Available in multiple pack sizes, including 25 g.