Learn More
IDEX Health & Science Upchurch 10 TO 32 FEMALE TO MALE LUER
Catalog No. NC1919471
Encompass_Preferred
Idex Threaded Luer PEEK Adapter, Natural, For Use With: To Female 10-32, Luer Adapter, Diameter Inner: 0.05 in., Temperature Max: 125 deg.C
Catalog No. NC1919471 Supplier IDEX Health & Science Upchurch Supplier No. P656
Idex Threaded Luer PEEK Adapter, Natural, For Use With: To Female 10-32, Luer Adapter, Diameter Inner: 0.05 in., Temperature Max: 125 deg.C