Learn More
TRIBIOSCIENCE INC 10% Trehalose Solution, 100 ML
SDP
Catalog No. NC3834047
Encompass
Trehalose dihydrate is a non-reducing disaccharide widely used in the formulation of biologic drugs as a stabilizing agent. It functions by protecting biomolecules such as proteins and antibodies from aggregation and degradation during manufacturing, purification, and storage. Due to its low bioburden and endotoxin levels, 10% Trehalose solution is especially suitable for biological applications.
Catalog No. NC3834047 Supplier TRIBIOSCIENCE INC Supplier No. TBS8078100ML
Trehalose dihydrate is a non-reducing disaccharide widely used in the formulation of biologic drugs as a stabilizing agent. It functions by protecting biomolecules such as proteins and antibodies from aggregation and degradation during manufacturing, purification, and storage. Due to its low bioburden and endotoxin levels, 10% Trehalose solution is especially suitable for biological applications.