Learn More
Teknova 10% Tween-20. 1000mL, Sterile.
SDP
Catalog No. 502387807
Encompass
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.
Catalog No. 50-238-7807 Supplier Teknova Supplier No. T0124
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.