Learn More
Bio Rad Laboratories 10% TWEEN 20
Catalog No. NC9744583
Encompass
Tween 20 is used as a blocking and wash buffer component in blotting. This 10% w/v Tween 20 is supplied as 1 L. (161-0781)
Catalog No. NC9744583 Supplier Bio Rad Laboratories Supplier No. 1610781
Tween 20 is used as a blocking and wash buffer component in blotting. This 10% w/v Tween 20 is supplied as 1 L. (161-0781)