Learn More
Seracare Life Sciences Inc Tween 20, 10%, nonionic detergent additive
Catalog No. 5067474
Encompass_Preferred
Tween 20 is a nonionic detergent additive for phosphate-buffer saline (PBS) and Western blotting wash solution that reduces non-specific binding and protein-protein interaction during the wash step in Western blotting procedures.
This item is not returnable. View return policy
Catalog No. 50-674-74 Supplier Seracare Life Sciences Inc Supplier No. 54600019
Tween 20 is a nonionic detergent additive for phosphate-buffer saline (PBS) and Western blotting wash solution that reduces non-specific binding and protein-protein interaction during the wash step in Western blotting procedures.