Learn More
Teknova 10% Tween-20 Solution. 100mL, Sterile.
SDP
Catalog No. 50843255
Encompass
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.
This item is not returnable. View return policy
Catalog No. 50-843-255 Supplier Teknova Supplier No. T0710
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.