Learn More
Teknova 10% Tween-20 Solution. 200mL, Sterile. 8 pack.
SDP
Catalog No. 502745982
Encompass
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.
Catalog No. 50-274-5982 Supplier Teknova Supplier No. T00258PK
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.