Learn More
Teknova 10% Tween-20 Solution, 500mL, Sterile. 2 Pack.
SDP
Catalog No. 502746026
Encompass
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.
Catalog No. 50-274-6026 Supplier Teknova Supplier No. 213382PK
Tween Solution is a nonionic detergent used for protein extraction and isolation of nuclei from mammalian cell lines. It is soluble in water and is widely used in biochemical applications such as solubilizing proteins, isolating nuclei from cells, and emulsifying and dispersing substances in medicinal and food products.