Learn More
Abcam Recombinant mouse Aurora A protein, 10UG
Supplier:  Abcam AB6032710UG
Encompass_Preferred
Recombinant mouse Aurora A protein is a Mouse Full Length protein, expressed in Baculovirus infected Sf9, with >90% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Aik, Airk1, Ark1, Aura, Ayk1, Btak, Iak1, Stk15, Stk6, Aurka, Aurora kinase A, Aurora 2, Aurora/IPL1-related kinase 1, Ipl1- and aurora-related kinase 1, Serine/threonine-protein kinase 15, Serine/threonine-protein kinase 6, Serine/threonine-protein kinase Ayk1, Serine/threonine-protein kinase aurora-A, ARK-1, Aurora-related kinase 1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2222298