Learn More
Gerstel GERSTEL 10 µL TriStar Syringe, Fixed Needle, 23s Gauge, 10 µL
SDP
Catalog No. NC1140433
Encompass
The GERSTEL 10 µL TriStar Syringe with Fixed Needle is designed for precise liquid handling, offering a 10 µL volume capacity and a robust 23s gauge fixed needle. Its reliable performance in various scientific workflows enhances sample introduction, ensuring direct and repeatable liquid transfer. Ideal for use with universal syringe holders and manual injection systems, this syringe differentiates itself with its fixed cone tip, making it a trustworthy option for analytical instrumentation.
Catalog No. NC1140433 Supplier Gerstel Supplier No. 02200005000
The GERSTEL 10 µL TriStar Syringe with Fixed Needle is designed for precise liquid handling, offering a 10 µL volume capacity and a robust 23s gauge fixed needle. Its reliable performance in various scientific workflows enhances sample introduction, ensuring direct and repeatable liquid transfer. Ideal for use with universal syringe holders and manual injection systems, this syringe differentiates itself with its fixed cone tip, making it a trustworthy option for analytical instrumentation.