Learn More
Waters Corp Syringe, 10 μL, Liquid, for GC-PAL 3 System
Catalog No. 502391492 Shop All Waters Corporation Products
Encompass_Preferred
This smart syringe is a replacement part designed for use with the GC-PAL 3 system. It offers a 10 μL volume capacity, ensuring precise liquid handling for various laboratory applications.
  • Designed for GC-PAL 3 system compatibility.
  • Features a 10 μL volume capacity.
  • Supplied as a single pack.
  • Recommended for annual replacement.
  • Categorized as a spare part kit.
Catalog No. 50-239-1492 Supplier Waters Corp Supplier No. 700012540
This smart syringe is a replacement part designed for use with the GC-PAL 3 system. It offers a 10 μL volume capacity, ensuring precise liquid handling for various laboratory applications.
  • Designed for GC-PAL 3 system compatibility.
  • Features a 10 μL volume capacity.
  • Supplied as a single pack.
  • Recommended for annual replacement.
  • Categorized as a spare part kit.