Learn More
USA Scientific Inc 10 ul TipOne RPT tip wafers, case of 10 refill packs
Supplier:  USA Scientific Inc 11613730
Encompass
TipOne RPT 10 ul ultra low retention pipet tip refill wafers, case of 10 refill packs (100 wafers of 96 tips; 9600 tips total)
Catalog No. NC1674960