Learn More
Medchemexpress LLC 10-Undecen-1-ol | 112-43-6 | 170.29 | 100 G
SDP
Catalog No. 5000448827
Encompass_Preferred
10-Undecen-1-ol, converted from ricinoleic acid, can be used as a comonomer for the introduction of functional groups. It is for research use only and not sold to patients. The compound is described as a liquid with a density of 0.85 g/cm³ and a colorless to light yellow appearance.
  • Converted from ricinoleic acid
  • Can be used as a comonomer for the introduction of functional groups
  • Intended for research use only
  • Liquid appearance
  • Colorless to light yellow appearance
  • Density of 0.85 g/cm³
Catalog No. 50-004-48827 Supplier Medchemexpress LLC Supplier No. HYW004298100G
10-Undecen-1-ol, converted from ricinoleic acid, can be used as a comonomer for the introduction of functional groups. It is for research use only and not sold to patients. The compound is described as a liquid with a density of 0.85 g/cm³ and a colorless to light yellow appearance.
  • Converted from ricinoleic acid
  • Can be used as a comonomer for the introduction of functional groups
  • Intended for research use only
  • Liquid appearance
  • Colorless to light yellow appearance
  • Density of 0.85 g/cm³