Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-Undecen-1-ol | 112-43-6 | 170.29 | 25 G
SDP

Catalog No. 5000448197
Encompass_Preferred

10-Undecen-1-ol is a compound converted from ricinoleic acid. It is primarily used as a comonomer for the introduction of functional groups and is intended for research purposes only. This product is supplied as a colorless to light yellow liquid.

  • Used as a comonomer for the introduction of functional groups
  • Converted from ricinoleic acid
  • For research use only
  • Appears as a colorless to light yellow liquid
  • Molecular formula: C11H22O
  • Molecular weight: 170.29
  • Purity: 99.6%

Catalog No. 50-004-48197 Supplier Medchemexpress LLC Supplier No. 25GHYW004298
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10-Undecen-1-ol is a compound converted from ricinoleic acid. It is primarily used as a comonomer for the introduction of functional groups and is intended for research purposes only. This product is supplied as a colorless to light yellow liquid.

  • Used as a comonomer for the introduction of functional groups
  • Converted from ricinoleic acid
  • For research use only
  • Appears as a colorless to light yellow liquid
  • Molecular formula: C11H22O
  • Molecular weight: 170.29
  • Purity: 99.6%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.