Learn More
Medchemexpress LLC 10-Undecen-1-ol | 112-43-6 | 170.29 | 50 G
SDP
Catalog No. 5000445490
Encompass_Preferred
10-Undecen-1-ol is derived from ricinoleic acid and serves as a comonomer for introducing functional groups. It can be used in cobalt-catalyzed step-growth polymerization with CO to produce aliphatic polyesters from renewable resources.
  • Can be used as a comonomer for the introduction of functional groups
  • Affords aliphatic polyesters based entirely on renewable resources
  • Colorless to light yellow liquid
  • Density of 0.85 g/cm³
Catalog No. 50-004-45490 Supplier Medchemexpress LLC Supplier No. HYW00429850G
10-Undecen-1-ol is derived from ricinoleic acid and serves as a comonomer for introducing functional groups. It can be used in cobalt-catalyzed step-growth polymerization with CO to produce aliphatic polyesters from renewable resources.
  • Can be used as a comonomer for the introduction of functional groups
  • Affords aliphatic polyesters based entirely on renewable resources
  • Colorless to light yellow liquid
  • Density of 0.85 g/cm³