Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 10-Undecen-1-ol | 112-43-6 | 170.29 | 500 G
SDP

Catalog No. 5000448828
Encompass_Preferred

10-Undecen-1-ol, converted from ricinoleic acid, can be used as a comonomer for the introduction of functional groups. In vitro, cobalt-catalyzed step growth polymerization of undec-10-en-1-ol and CO affords aliphatic polyesters (Mn 104 g/mol) based entirely on renewable resources.

  • Purity: 99.56%
  • Molecular weight: 170.29
  • Formula: C11H22O
  • Appearance as a liquid
  • Colorless to light yellow
  • Used as a comonomer for functional group introduction
  • Supports cobalt-catalyzed step growth polymerization to form aliphatic polyesters from renewable resources

Catalog No. 50-004-48828 Supplier Medchemexpress LLC Supplier No. HYW004298500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10-Undecen-1-ol, converted from ricinoleic acid, can be used as a comonomer for the introduction of functional groups. In vitro, cobalt-catalyzed step growth polymerization of undec-10-en-1-ol and CO affords aliphatic polyesters (Mn 104 g/mol) based entirely on renewable resources.

  • Purity: 99.56%
  • Molecular weight: 170.29
  • Formula: C11H22O
  • Appearance as a liquid
  • Colorless to light yellow
  • Used as a comonomer for functional group introduction
  • Supports cobalt-catalyzed step growth polymerization to form aliphatic polyesters from renewable resources

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.