Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 10-Undecen-1-ol

Catalog No. U2008100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U2008100G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U2008100G Supplier Sigma Aldrich Supplier No. U2008100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

10-Undecen-1-ol

Specifications

CAS 112-43-6
Density 0.85 g/mL (at 25°C (literature))
Boiling Point 132°C to 133°C (15 mmHg), 245°C to 248°C (lit.)
Molecular Formula C11H22O
Refractive Index n20/D 1.448-1.453; n20/D 1.45 (literature)
Linear Formula CH2=CH(CH2)9 OH
MDL Number MFCD00004750
Quantity 25 g
Synonym 11-Hydroxy-1-undecene; NSC 66157; Undecylenyl alcohol
Molecular Weight (g/mol) 170.29
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.