Learn More
Medchemexpress LLC HY-W004298 500mg Medchemexpress, 10-Undecen-1-ol CAS:112-43-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW004298500MG
Encompass_Preferred
Medchemexpress, HY-W004298 500mg 10-Undecen-1-ol CAS:112-43-6 10-Undecen-1-ol, converted from ricinoleic acid, can be used as a comonomer for the introduction of functional groups. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3219